Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004072.2 | NP_001004072.2 | 9 - 10 | 934 | 134 | |
GenBank mRNA | BC079369.1 | - | 9 - 10 | 926 | 134 | |
BC098897.1 | - | 9 - 10 | 934 | 7103543 | 134 | |
Z12158.1 | - | 9 - 10 | 953 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |