Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rogdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024864.1 | NP_001020035.1 | 4 - 5 | 447 | 119 | |
GenBank mRNA | BC098001.1 | - | 4 - 5 | 447 | 7453517 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |