Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012573.3 | NP_036705.3 | 11 - 12 | 2575 | 102 | |
GenBank mRNA | D13211.1 | - | 10 - 11 | 2368 | 102 | |
M91561.1 | - | 11 - 12 | 2575 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |