Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038595.1 | NP_001033684.1 | 1 - 2 | 210 | 73 | |
NM_001038596.1 | NP_001033685.1 | 1 - 2 | 210 | 73 | ||
XM_006245817.2 | XP_006245879.1 | 1 - 2 | 250 | 73 | ||
GenBank mRNA | AF516338.1 | - | 1 - 2 | 210 | 73 | |
AY077460.1 | - | 1 - 2 | 210 | 73 | ||
AY264842.1 | - | 1 - 2 | 210 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |