Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038595.1 | NP_001033684.1 | 2 - 3 | 343 | 101 | |
NM_001038596.1 | NP_001033685.1 | 2 - 3 | 343 | 101 | ||
XM_006245817.2 | XP_006245879.1 | 2 - 3 | 383 | 101 | ||
GenBank mRNA | AF516338.1 | - | 2 - 3 | 343 | 101 | |
AY077460.1 | - | 2 - 3 | 343 | 101 | ||
AY264842.1 | - | 2 - 3 | 343 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |