Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108264.1 | NP_001101734.1 | 2 - 3 | 292 | 127 | |
XM_006245802.2 | XP_006245864.1 | 2 - 3 | 328 | 127 | ||
XM_008767491.1 | XP_008765713.1 | 2 - 3 | 134 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |