Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014225.1 | NP_001014247.1 | 7 - 8 | 1050 | 125 | |
XM_006245832.2 | XP_006245894.1 | 7 - 8 | 813 | 125 | ||
XM_008767512.1 | XP_008765734.1 | 7 - 8 | 746 | 125 | ||
XM_008767513.1 | XP_008765735.1 | 7 - 8 | 1108 | 125 | ||
XM_008767514.1 | XP_008765736.1 | 6 - 7 | 650 | 125 | ||
XM_008767515.1 | XP_008765737.1 | 6 - 7 | 654 | 125 | ||
XM_008767516.1 | XP_008765738.1 | 6 - 7 | 652 | 125 | ||
XM_008767517.1 | XP_008765739.1 | 5 - 6 | 515 | 125 | ||
XM_008767518.1 | XP_008765740.1 | 5 - 6 | 572 | 125 | ||
GenBank mRNA | BC079022.1 | - | 7 - 8 | 1050 | 7113868 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |