Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053661.1 | NP_446113.1 | 1 - 2 | 56 | 97 | |
XM_006236610.2 | XP_006236672.1 | 2 - 3 | 620 | 97 | ||
XM_008762969.1 | XP_008761191.1 | 2 - 3 | 520 | 97 | ||
XM_008762970.1 | XP_008761192.1 | 2 - 3 | 495 | 97 | ||
GenBank mRNA | FM031435.1 | - | 2 - 3 | 74 | 97 | |
FQ223368.1 | - | 2 - 3 | 94 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |