Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nagk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037768.1 | NP_001032857.1 | 8 - 9 | 798 | 83 | |
GenBank mRNA | BC107647.1 | - | 8 - 9 | 798 | 7930414 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |