Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012735.2 | NP_036867.1 | 2 - 3 | 421 | 79 | |
GenBank mRNA | AF027179.1 | - | 2 - 3 | 225 | 79 | |
M68971.1 | - | 2 - 3 | 422 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |