Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039686.1 | NP_001034775.1 | 5 - 6 | 505 | 132 | |
XM_008763064.1 | XP_008761286.1 | 4 - 5 | 2033 | 132 | ||
XM_008763065.1 | XP_008761287.1 | 8 - 9 | 989 | 132 | ||
GenBank mRNA | AY943907.1 | - | 5 - 6 | 505 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |