Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013087.1 | NP_001013105.1 | 7 - 8 | 761 | 117 | |
XM_006236667.2 | XP_006236729.1 | 7 - 8 | 761 | 117 | ||
XM_006236669.1 | XP_006236731.1 | 6 - 7 | 713 | 117 | ||
XM_008762978.1 | XP_008761200.1 | 6 - 7 | 723 | 117 | ||
GenBank mRNA | BC083623.1 | - | 7 - 8 | 761 | 7134106 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |