Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053879.1 | NP_446331.1 | 20 - 21 | 2816 | 104 | |
GenBank mRNA | U35371.1 | - | 20 - 21 | 2816 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |