Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749809.3 | XP_003749857.1 | 2 - 3 | 1301 | 149 | |
XM_006237062.2 | XP_006237124.1 | 2 - 3 | 1379 | 149 | ||
XM_006237064.2 | XP_006237126.1 | 2 - 3 | 1224 | 149 | ||
XM_008763200.1 | XP_008761422.1 | 1 - 2 | 1223 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |