Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mitf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF029886.1 | - | 1 - 2 | 85 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |