Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_232051.8 | XP_232051.4 | 3 - 4 | 508 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |