Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109398.1 | NP_001102868.1 | 4 - 5 | 639 | 66 | |
GenBank mRNA | FQ234580.1 | - | 5 - 6 | 691 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |