Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109407.1 | NP_001102877.1 | 8 - 9 | 1566 | 77 | |
XM_006236717.2 | XP_006236779.1 | 8 - 9 | 2056 | 77 | ||
XM_008763030.1 | XP_008761252.1 | 7 - 8 | 2241 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |