Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109407.1 | NP_001102877.1 | 9 - 10 | 1686 | 135 | |
XM_006236717.2 | XP_006236779.1 | 9 - 10 | 2176 | 135 | ||
XM_008763030.1 | XP_008761252.1 | 8 - 9 | 2361 | 135 |
Gene Symbol: LOC103692182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763119.1 | XP_008761341.1 | 242 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |