Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009502.1 | NP_001009502.1 | 6 - 7 | 866 | 86 | |
XM_008763144.1 | XP_008761366.1 | 3 - 4 | 540 | 86 | ||
XM_008763145.1 | XP_008761367.1 | 4 - 5 | 452 | 86 | ||
XM_008763146.1 | XP_008761368.1 | 2 - 3 | 332 | 86 | ||
XM_008763147.1 | XP_008761369.1 | 3 - 4 | 597 | 86 | ||
GenBank mRNA | AJ868236.1 | - | 6 - 7 | 866 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |