Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025716.1 | NP_001020887.1 | 7 - 8 | 983 | 77 | |
GenBank mRNA | BC099128.1 | - | 7 - 8 | 983 | 7459593 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |