Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108896.1 | NP_001102366.1 | 3 - 4 | 285 | 68 | |
GenBank mRNA | FQ228966.1 | - | 3 - 4 | 203 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |