Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255732.2 | XP_006255794.1 | 3 - 4 | 815 | 154 | |
XM_344780.7 | XP_344781.5 | 3 - 4 | 815 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |