Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037094.1 | NP_001032171.1 | 8 - 9 | 888 | 105 | |
GenBank mRNA | BC079393.1 | - | 8 - 9 | 938 | 105 | |
BC089862.1 | - | 8 - 9 | 886 | 105 | ||
BC107464.1 | - | 8 - 9 | 888 | 7454431 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |