Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057148.2 | NP_476489.1 | 1 - 2 | 41 | 74 | |
GenBank mRNA | BC081745.1 | - | 1 - 2 | 41 | 7121592 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |