Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767380.1 | XP_008765602.1 | 1 - 2 | 1155 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |