Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750733.2 | XP_003750781.1 | 5 - 6 | 1322 | 73 | |
GenBank mRNA | FQ216933.1 | - | 5 - 6 | 1290 | 73 | |
FQ216966.1 | - | 5 - 6 | 1322 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |