Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco4c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002024.1 | NP_001002024.1 | 1 - 2 | 340 | 92 | |
GenBank mRNA | AF454761.1 | - | 1 - 2 | 340 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |