Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127683.1 | NP_001121155.1 | 6 - 7 | 711 | 117 | |
XM_008772826.1 | XP_008771048.1 | 7 - 8 | 947 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |