Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U40064.1 | - | 1 - 2 | 92 | 154 | |
U75918.1 | - | 1 - 2 | 84 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |