Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017299.2 | NP_058995.2 | 1 - 2 | 201 | 66 | |
GenBank mRNA | AF099009.1 | - | 1 - 2 | 210 | 66 | |
AF099010.1 | - | 1 - 2 | 210 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |