Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tapbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033098.2 | NP_149089.1 | 4 - 5 | 895 | 126 | |
GenBank mRNA | AJ400732.1 | - | 4 - 5 | 884 | 126 | |
BC062007.1 | - | 4 - 5 | 886 | 5599850 | 126 | |
KC222917.1 | - | 4 - 5 | 872 | 126 | ||
KC222918.1 | - | 4 - 5 | 872 | 126 | ||
KC222919.1 | - | 4 - 5 | 872 | 126 | ||
KC222920.1 | - | 4 - 5 | 872 | 126 | ||
KC222921.1 | - | 4 - 5 | 872 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |