Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012796.2 | NP_036928.1 | 3 - 4 | 487 | 85 | |
GenBank mRNA | BC061856.1 | - | 3 - 4 | 491 | 5623709 | 85 |
CK473092.1 | - | 3 - 4 | 487 | 85 | ||
D10026.1 | - | 3 - 4 | 475 | 85 | ||
FJ179409.1 | - | 3 - 4 | 353 | 85 |
Gene Symbol: LOC103694879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773848.1 | XP_008772070.1 | 1 - 2 | 557 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |