Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032056.3 | NP_114445.2 | 10 - 11 | 1886 | 78 | |
GenBank mRNA | BC091140.1 | - | 9 - 10 | 1819 | 7381526 | 78 |
KC222910.1 | - | 9 - 10 | 1797 | 78 | ||
X75305.1 | - | 10 - 11 | 1833 | 78 |
Gene Symbol: LOC103689996 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772777.1 | XP_008770999.1 | 10 - 11 | 1938 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |