Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107618.1 | NP_001101088.1 | 6 - 7 | 1343 | 67 | |
XM_006256207.2 | XP_006256269.1 | 6 - 7 | 1915 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |