Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212538.1 | NP_997703.1 | 5 - 6 | 652 | 64 | |
XM_006256117.2 | XP_006256179.1 | 5 - 6 | 906 | 64 | ||
XM_008772723.1 | XP_008770945.1 | 5 - 6 | 907 | 64 | ||
GenBank mRNA | BC083843.1 | - | 5 - 6 | 593 | 7114670 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |