Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039032.1 | NP_001034121.1 | 4 - 5 | 557 | 63 | |
GenBank mRNA | BC098775.1 | - | 4 - 5 | 557 | 7373953 | 63 |
Gene Symbol: NEWGENE_1305341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772790.1 | XP_008771012.1 | 2 - 3 | 240 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |