Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008885.2 | NP_001008885.1 | 3 - 4 | 948 | 96 | |
NM_001164744.1 | NP_001158216.1 | 4 - 5 | 689 | 96 | ||
GenBank mRNA | BC079141.1 | - | 3 - 4 | 948 | 7127902 | 96 |
FQ212335.1 | - | 4 - 5 | 656 | 96 | ||
FQ212833.1 | - | 3 - 4 | 976 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |