Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008885.2 | NP_001008885.1 | 5 - 6 | 1262 | 162 | |
NM_001164744.1 | NP_001158216.1 | 6 - 7 | 1003 | 162 | ||
GenBank mRNA | BC079141.1 | - | 5 - 6 | 1262 | 7127902 | 162 |
FQ212335.1 | - | 6 - 7 | 970 | 162 | ||
FQ212833.1 | - | 5 - 6 | 1289 | 161 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |