Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007695.1 | NP_001007696.1 | 8 - 9 | 1362 | 83 | |
XM_006256200.2 | XP_006256262.1 | 8 - 9 | 1221 | 83 | ||
GenBank mRNA | BC083899.1 | - | 8 - 9 | 1362 | 7133508 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |