Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 14 - 15 | 5688 | 70 | |
XM_006256359.2 | XP_006256421.1 | 14 - 15 | 5742 | 70 | ||
XM_006256360.2 | XP_006256422.1 | 13 - 14 | 5538 | 70 | ||
XM_006256361.2 | XP_006256423.1 | 14 - 15 | 5765 | 70 | ||
XM_006256362.2 | XP_006256424.1 | 14 - 15 | 5657 | 70 | ||
XM_006256363.1 | XP_006256425.1 | 10 - 11 | 5099 | 70 | ||
XM_006256364.1 | XP_006256426.1 | 14 - 15 | 5651 | 70 | ||
XM_006256365.1 | XP_006256427.1 | 9 - 10 | 4957 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |