Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 18 - 19 | 6527 | 87 | |
XM_006256359.2 | XP_006256421.1 | 18 - 19 | 6581 | 87 | ||
XM_006256360.2 | XP_006256422.1 | 17 - 18 | 6377 | 87 | ||
XM_006256361.2 | XP_006256423.1 | 18 - 19 | 6604 | 87 | ||
XM_006256362.2 | XP_006256424.1 | 18 - 19 | 6496 | 87 | ||
XM_006256363.1 | XP_006256425.1 | 14 - 15 | 5938 | 87 | ||
XM_006256364.1 | XP_006256426.1 | 18 - 19 | 6490 | 87 | ||
XM_006256365.1 | XP_006256427.1 | 13 - 14 | 5796 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |