Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109589.2 | NP_001103059.1 | 10 - 11 | 802 | 68 | |
XM_006256342.2 | XP_006256404.1 | 10 - 11 | 1232 | 68 | ||
GenBank mRNA | BC167029.1 | - | 10 - 11 | 802 | 8368465 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |