Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107642.1 | NP_001101112.1 | 11 - 12 | 1159 | 77 | |
XM_006256217.2 | XP_006256279.1 | 13 - 14 | 1419 | 77 | ||
XM_006256218.2 | XP_006256280.1 | 12 - 13 | 1413 | 77 | ||
XM_006256219.2 | XP_006256281.1 | 13 - 14 | 1419 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons |