Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107613.3 | NP_001101083.1 | 9 - 10 | 1412 | 74 | |
XM_006256193.2 | XP_006256255.1 | 10 - 11 | 1487 | 74 | ||
XM_006256195.2 | XP_006256257.1 | 9 - 10 | 1424 | 74 | ||
XM_006256196.2 | XP_006256258.1 | 10 - 11 | 1487 | 74 | ||
XM_006256197.2 | XP_006256259.1 | 9 - 10 | 1424 | 74 | ||
XM_008772813.1 | XP_008771035.1 | 10 - 11 | 1487 | 74 | ||
XM_008772814.1 | XP_008771036.1 | 2 - 3 | 1348 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |