Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100833.1 | NP_001094303.1 | 5 - 6 | 653 | 95 | |
XM_006256184.1 | XP_006256246.1 | 5 - 6 | 675 | 95 | ||
GenBank mRNA | BC158661.1 | - | 5 - 6 | 653 | 8369713 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |