Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212549.2 | NP_997714.2 | 1 - 2 | 145 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |