Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212549.2 | NP_997714.2 | 2 - 3 | 279 | 69 | |
GenBank mRNA | AB568263.1 | - | 1 - 2 | 63 | 69 | |
BC098635.1 | - | 2 - 3 | 223 | 7098264 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |