Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106380.1 | NP_001099850.1 | 2 - 3 | 154 | 93 | |
GenBank mRNA | BC166733.1 | - | 2 - 3 | 87 | 7936238 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |