Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pofut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107621.2 | NP_001101091.2 | 4 - 5 | 705 | 84 | |
XM_006256299.2 | XP_006256361.1 | 4 - 5 | 1300 | 84 | ||
GenBank mRNA | CV103570.1 | - | 3 - 4 | 437 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |